Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(4-(2-tert-butylphenyl)sulfonylphenyl)-2,3,4-trihydroxy-5-(2-isopropylbenzyl)benzamide | 877877-35-5 | 99.5% | 1 ML
SDP

Catalog No. 5000233222
Encompass_Preferred

TW-37 is a small-molecule BH3 mimetic that inhibits anti-apoptotic Bcl-2 family proteins and is used in apoptosis and oncology research. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for research use only.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Potent inhibition of Bcl-2 family proteins with Ki values: Mcl-1 260 nM; Bcl-2 290 nM; Bcl-xL 1,110 nM.
  • Molecular formula: C33H35NO6S; molecular weight: 573.7 g·mol⁻1.
  • High purity (99.5%) suitable for research applications.
  • Intended for in vitro research use; not for clinical or therapeutic use.

Catalog No. 50-002-33222 Supplier Medchemexpress LLC Supplier No. HY1202010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TW-37 is a small-molecule BH3 mimetic that inhibits anti-apoptotic Bcl-2 family proteins and is used in apoptosis and oncology research. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for research use only.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Potent inhibition of Bcl-2 family proteins with Ki values: Mcl-1 260 nM; Bcl-2 290 nM; Bcl-xL 1,110 nM.
  • Molecular formula: C33H35NO6S; molecular weight: 573.7 g·mol⁻1.
  • High purity (99.5%) suitable for research applications.
  • Intended for in vitro research use; not for clinical or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.