Learn More
Medchemexpress LLC N-(4-((3-chloro-4-fluorophenyl)amino)pyrido[3,4-d]pyrimidin-6-yl)-N'-(1-(dimethylamino)propan-2-yl)a | 848133-17-5 | MFCD26792555 | 99.2% | 574.05 | C31H29ClFN5O3 | 5 MG
SDP
Catalog No. 5000245116
Encompass_Preferred
HKI-357 is an irreversible small-molecule inhibitor of EGFR and ERBB2 kinases. It inhibits EGFR autophosphorylation at Y1068 and reduces downstream AKT and MAPK phosphorylation, showing potent activity in cell signaling and preclinical cancer models. Supplied as a solid research reagent for in vitro and preclinical studies.
  • Irreversible dual inhibitor of EGFR and ERBB2
  • Potent activity: EGFR 34 nM, ERBB2 33 nM
  • Suppresses EGFR Y1068 autophosphorylation and downstream signaling
  • Supplied as a solid, >99% purity (batch-dependent)
  • Suitable for kinase signaling and preclinical cancer research
Catalog No. 50-002-45116 Supplier Medchemexpress LLC Supplier No. HY1034435MG
HKI-357 is an irreversible small-molecule inhibitor of EGFR and ERBB2 kinases. It inhibits EGFR autophosphorylation at Y1068 and reduces downstream AKT and MAPK phosphorylation, showing potent activity in cell signaling and preclinical cancer models. Supplied as a solid research reagent for in vitro and preclinical studies.
  • Irreversible dual inhibitor of EGFR and ERBB2
  • Potent activity: EGFR 34 nM, ERBB2 33 nM
  • Suppresses EGFR Y1068 autophosphorylation and downstream signaling
  • Supplied as a solid, >99% purity (batch-dependent)
  • Suitable for kinase signaling and preclinical cancer research