Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[4-[[[(3R,4R)-1-[(2S)-2-amino-1-oxopropyl]-3-methyl-4-piperidinyl]amino]sulfonyl]-1-naphthalenyl] | 850330-18-6 | 99.8% | 508.63 g·mol^-1 | C27H32N4O4S | 25MG
SDP

Catalog No. 5000238301
Encompass_Preferred

ML604086 is a selective small-molecule inhibitor of the chemokine receptor CCR8 developed for laboratory research. It inhibits CCL1 binding to CCR8, blocks CCL1-mediated chemotaxis, and increases intracellular Ca2+ in CCR8-expressing cells, with reported micromolar activity in vitro.

  • Selective CCR8 small-molecule inhibitor for research use.
  • Inhibits CCL1 binding to CCR8 and blocks chemotaxis.
  • Increases intracellular Ca2+ concentrations in CCR8-expressing cells.
  • Reported in vitro IC50 ~1.0-1.3 μM in CCR8 assays.
  • Chemical formula C27H32N4O4S; molecular weight 508.63 g·mol⁻¹.
  • Purity listed as 99.78% (supplier assay).
  • Available as powder and as 10 mM solution in DMSO, with multiple pack sizes including 25 mg.
  • Intended for research use only; not for human use.

Catalog No. 50-002-38301 Supplier Medchemexpress LLC Supplier No. HY12441625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ML604086 is a selective small-molecule inhibitor of the chemokine receptor CCR8 developed for laboratory research. It inhibits CCL1 binding to CCR8, blocks CCL1-mediated chemotaxis, and increases intracellular Ca2+ in CCR8-expressing cells, with reported micromolar activity in vitro.

  • Selective CCR8 small-molecule inhibitor for research use.
  • Inhibits CCL1 binding to CCR8 and blocks chemotaxis.
  • Increases intracellular Ca2+ concentrations in CCR8-expressing cells.
  • Reported in vitro IC50 ~1.0-1.3 μM in CCR8 assays.
  • Chemical formula C27H32N4O4S; molecular weight 508.63 g·mol⁻¹.
  • Purity listed as 99.78% (supplier assay).
  • Available as powder and as 10 mM solution in DMSO, with multiple pack sizes including 25 mg.
  • Intended for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.