Learn More
Medchemexpress LLC N-(4-(4-(2-(diethylamino)-2-oxo-1-phenylethyl)piperazinyl)-3-fluorophenyl)-2-(3-pyridyl)benzamide | 1094873-14-9 | MFCD18782744 | 98.0% | C34H36FN5O2 | 10MG
SDP
Catalog No. 5000258509
Encompass_Preferred
JNJ-31020028 is a selective, brain-penetrant antagonist of the neuropeptide Y Y2 receptor used in neuroscience research. It demonstrates high potency in human and rodent Y2 assays and is supplied as a well-characterized, high-purity research compound with documented solubility and storage recommendations for powder and solution forms.
  • High purity suitable for research applications.
  • Brain-penetrant antagonist of neuropeptide Y Y2 receptor.
  • Demonstrated potency in human and rodent assays.
  • Compatible with common in vitro and in vivo formulations.
  • Documented solubility and storage conditions for reproducibility.
This item is not returnable. View return policy
Catalog No. 50-002-58509 Supplier Medchemexpress LLC Supplier No. HY1445010MG
JNJ-31020028 is a selective, brain-penetrant antagonist of the neuropeptide Y Y2 receptor used in neuroscience research. It demonstrates high potency in human and rodent Y2 assays and is supplied as a well-characterized, high-purity research compound with documented solubility and storage recommendations for powder and solution forms.
  • High purity suitable for research applications.
  • Brain-penetrant antagonist of neuropeptide Y Y2 receptor.
  • Demonstrated potency in human and rodent assays.
  • Compatible with common in vitro and in vivo formulations.
  • Documented solubility and storage conditions for reproducibility.