Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(4-(4-(2-methoxyethoxy)phenyl)thiazol-2-yl)isoxazole-5-carboxamide | 2375779-31-8 | 99.3% | 345.37 g/mol | C16H15N3O4S | 10 MG
SDP

Catalog No. 5000285431
Encompass_Preferred

GP130 modulator-1 (compound A33) is a small-molecule modulator of the gp130 signaling pathway intended for research use in studies of inflammatory and degenerative disorders.

  • Small-molecule gp130 signaling pathway modulator for research use.
  • High purity: 99.3%.
  • Molecular weight 345.37 g/mol.
  • Chemical formula C16H15N3O4S.
  • Available in multiple pack sizes, including 10 mg.
  • Supplied as a light yellow to yellow solid for laboratory handling and storage.

Catalog No. 50-002-85431 Supplier Medchemexpress LLC Supplier No. HY15974210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GP130 modulator-1 (compound A33) is a small-molecule modulator of the gp130 signaling pathway intended for research use in studies of inflammatory and degenerative disorders.

  • Small-molecule gp130 signaling pathway modulator for research use.
  • High purity: 99.3%.
  • Molecular weight 345.37 g/mol.
  • Chemical formula C16H15N3O4S.
  • Available in multiple pack sizes, including 10 mg.
  • Supplied as a light yellow to yellow solid for laboratory handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.