Learn More
Medchemexpress LLC N-[4-(4-amino-2-ethyl-1H-imidazo[4,5-c]quinolin-1-yl)butyl]methanesulfonamide | 532959-63-0 | MFCD30533308 | 99.2% | 361.5 g/mol | C17H23N5O2S | 100 MG
SDP
Catalog No. 5000215775
Encompass_Preferred
PF-4878691 is a small-molecule Toll-like receptor 7 (TLR7) agonist used in preclinical and research settings to activate innate immune signaling. It induces type I interferon responses, has reported antiviral efficacy against hepatitis C virus, and is utilized in studies of cancer immunology and TLR7 pathway modulation.
  • Potent TLR7 agonist activity in cellular and ex vivo assays.
  • Induces type I interferon and downstream antiviral signaling.
  • Demonstrated activity in antiviral and cancer immunology research.
  • Well characterized chemically (CAS 532959-63-0; molecular weight 361.5 g/mol).
  • Available from research suppliers in defined purity for experimental use.
Catalog No. 50-002-15775 Supplier Medchemexpress LLC Supplier No. HY100176100MG
PF-4878691 is a small-molecule Toll-like receptor 7 (TLR7) agonist used in preclinical and research settings to activate innate immune signaling. It induces type I interferon responses, has reported antiviral efficacy against hepatitis C virus, and is utilized in studies of cancer immunology and TLR7 pathway modulation.
  • Potent TLR7 agonist activity in cellular and ex vivo assays.
  • Induces type I interferon and downstream antiviral signaling.
  • Demonstrated activity in antiviral and cancer immunology research.
  • Well characterized chemically (CAS 532959-63-0; molecular weight 361.5 g/mol).
  • Available from research suppliers in defined purity for experimental use.