Learn More
Medchemexpress LLC N-[4-(4-amino-2-ethylimidazo[4,5-c]quinolin-1-yl)butyl]methanesulfonamide | 532959-63-0 | MFCD30533308 | 99.9% | 361.5 g/mol | C17H23N5O2S | 25 MG
SDP
Catalog No. 5000215581
Encompass_Preferred
PF-4878691 is an orally active small-molecule toll-like receptor 7 (TLR7) agonist used in preclinical research to stimulate innate immune responses. It has reported antiviral activity against hepatitis C virus and is applied in oncology and infectious disease research models.
  • Orally active TLR7 agonist.
  • Stimulates innate immune responses, including interferon production.
  • Demonstrated antiviral activity against hepatitis C virus.
  • Suitable for preclinical in vitro and in vivo research.
  • High chemical purity for research applications.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-15581 Supplier Medchemexpress LLC Supplier No. HY10017625MG
PF-4878691 is an orally active small-molecule toll-like receptor 7 (TLR7) agonist used in preclinical research to stimulate innate immune responses. It has reported antiviral activity against hepatitis C virus and is applied in oncology and infectious disease research models.
  • Orally active TLR7 agonist.
  • Stimulates innate immune responses, including interferon production.
  • Demonstrated antiviral activity against hepatitis C virus.
  • Suitable for preclinical in vitro and in vivo research.
  • High chemical purity for research applications.
  • For research use only; not for human or clinical use.