Learn More
Medchemexpress LLC N-[4-(4-chlorophenyl)sulfonyl-4-(2,5-difluorophenyl)cyclohexyl]-1,1,1-trifluoromethanesulfonamide | 677772-84-8 | MFCD09970622 | >98.0% | C19H17ClF5NO4S2 | 10MG
SDP
Catalog No. 5000259515
Encompass_Preferred
MRK-560 is a PSEN1-selective γ-secretase inhibitor used as a research probe for amyloid precursor protein processing. It is orally active and brain-penetrant, and has been shown in preclinical studies to reduce amyloid-β peptide levels in brain and cerebrospinal fluid. The compound is supplied as a solid powder with documented storage and solubility recommendations for in vitro and in vivo applications.
  • Selective inhibition of PSEN1-containing γ-secretase complexes.
  • Orally active and brain-penetrant for central nervous system studies.
  • Reduces amyloid-β peptide levels in preclinical models.
  • High solubility in DMSO and multiple documented in vivo formulation options.
  • Supplied as a powder with recommended cold-storage conditions for stability.
  • Well-characterized chemical identity with published molecular and structural data.
Catalog No. 50-002-59515 Supplier Medchemexpress LLC Supplier No. HY1417410MG
MRK-560 is a PSEN1-selective γ-secretase inhibitor used as a research probe for amyloid precursor protein processing. It is orally active and brain-penetrant, and has been shown in preclinical studies to reduce amyloid-β peptide levels in brain and cerebrospinal fluid. The compound is supplied as a solid powder with documented storage and solubility recommendations for in vitro and in vivo applications.
  • Selective inhibition of PSEN1-containing γ-secretase complexes.
  • Orally active and brain-penetrant for central nervous system studies.
  • Reduces amyloid-β peptide levels in preclinical models.
  • High solubility in DMSO and multiple documented in vivo formulation options.
  • Supplied as a powder with recommended cold-storage conditions for stability.
  • Well-characterized chemical identity with published molecular and structural data.