Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]-4-[(2-morpholin-4-yl-2-oxoethyl)amino]benzamide | 2073059-82-0 | 98.7% | 1 ML
SDP

Catalog No. 5000231299
Encompass_Preferred

SBI-993 is a small-molecule analog of SBI-477 that stimulates insulin signaling by deactivating the transcription factor MondoA. It is supplied as solid quantities and as a 10 mM solution in DMSO for research use.

  • Stimulates insulin signaling via MondoA deactivation.
  • Available as solid quantities (5, 10, 25, 50, 100 mg) and as a 10 mM DMSO solution (1 mL).
  • Molecular formula C23H24N4O4S and molecular weight 452.53 g/mol.
  • High purity suitable for biochemical assays (98.7%).
  • Soluble in DMSO for preparation of concentrated stock solutions.

Catalog No. 50-002-31299 Supplier Medchemexpress LLC Supplier No. HY12268210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SBI-993 is a small-molecule analog of SBI-477 that stimulates insulin signaling by deactivating the transcription factor MondoA. It is supplied as solid quantities and as a 10 mM solution in DMSO for research use.

  • Stimulates insulin signaling via MondoA deactivation.
  • Available as solid quantities (5, 10, 25, 50, 100 mg) and as a 10 mM DMSO solution (1 mL).
  • Molecular formula C23H24N4O4S and molecular weight 452.53 g/mol.
  • High purity suitable for biochemical assays (98.7%).
  • Soluble in DMSO for preparation of concentrated stock solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.