Learn More
Medchemexpress LLC N-[[4-(4-methylpiperazin-1-yl)phenyl]methyl]-1,2-oxazole-5-carboxamide | 883944-52-3 | 98.4% | C16H20N4O2 | 10MG
SDP
Catalog No. 5000260107
Encompass_Preferred
SSAA09E2 is a small-molecule inhibitor of SARS-CoV replication that interferes with early interactions between the viral spike (S) protein and the ACE2 receptor. It is supplied for laboratory research and is provided with structural identifiers and purity data to support assay development and compound management.
  • Inhibits SARS-CoV replication by blocking S protein-ACE2 interactions.
  • High purity (≈98.4%) suitable for biochemical and cell-based assays.
  • Available as solid quantities and as a DMSO solution for screening.
  • Includes molecular formula and SMILES for computational workflows.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-60107 Supplier Medchemexpress LLC Supplier No. HY13806710MG
SSAA09E2 is a small-molecule inhibitor of SARS-CoV replication that interferes with early interactions between the viral spike (S) protein and the ACE2 receptor. It is supplied for laboratory research and is provided with structural identifiers and purity data to support assay development and compound management.
  • Inhibits SARS-CoV replication by blocking S protein-ACE2 interactions.
  • High purity (≈98.4%) suitable for biochemical and cell-based assays.
  • Available as solid quantities and as a DMSO solution for screening.
  • Includes molecular formula and SMILES for computational workflows.
  • Intended for research use only; not for human or diagnostic use.