Learn More
Medchemexpress LLC N-[4-[(4R)-4-methyl-6-oxo-4,5-dihydro-1H-pyridazin-3-yl]phenyl]acetamide | 220246-81-1 | MFCD09952125 | 99.0% | C13H15N3O2 | 10MG
SDP
Catalog No. 5000258422
Encompass_Preferred
OR-1896 is the active, long-lived metabolite of levosimendan with calcium-sensitizing and phosphodiesterase III (PDE III) inhibitory activity. It is used in pharmacological and in vivo research to study cardioprotective effects, myocardial contractility, and cardiac remodeling.
  • Active metabolite with calcium-sensitizing and PDE III inhibitory properties.
  • Demonstrated cardioprotective effects in preclinical studies.
  • High solubility in DMSO for in vitro applications.
  • Formulatable for in vivo dosing using several vehicle systems.
  • High purity suitable for research use.
  • Stable as a powder when stored at low temperatures.
Catalog No. 50-002-58422 Supplier Medchemexpress LLC Supplier No. HY13574610MG
OR-1896 is the active, long-lived metabolite of levosimendan with calcium-sensitizing and phosphodiesterase III (PDE III) inhibitory activity. It is used in pharmacological and in vivo research to study cardioprotective effects, myocardial contractility, and cardiac remodeling.
  • Active metabolite with calcium-sensitizing and PDE III inhibitory properties.
  • Demonstrated cardioprotective effects in preclinical studies.
  • High solubility in DMSO for in vitro applications.
  • Formulatable for in vivo dosing using several vehicle systems.
  • High purity suitable for research use.
  • Stable as a powder when stored at low temperatures.