Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[4-[6-[4-(trifluoromethyl)phenyl]pyrimidin-4-yl]oxy-1,3-benzothiazol-2-yl]acetamide | 659730-32-2 | MFCD14584859 | 99.2% | 430.40 | C20H13F3N4O2S | 25 MG
SDP

Catalog No. 5000244040
Encompass_Preferred

AMG 517 is a potent, selective small-molecule antagonist of the vanilloid receptor TRPV1 for research use. It inhibits TRPV1-mediated calcium influx in cell-based and neuronal assays with low-nanomolar IC50 values, and is supplied as a high-purity solid suitable for pharmacology and biochemical studies.

  • High purity solid suitable for in vitro research.
  • Low-nanomolar TRPV1 potency (IC50 ≈ 0.5-0.9 nM).
  • Molecular weight 430.40, formula C20H13F3N4O2S.
  • Supplied with data sheet, certificate of analysis, and safety data sheet.
  • Soluble in DMSO for preparation of stock solutions.
  • Available in multiple pack sizes for flexibility in study scale.

Catalog No. 50-002-44040 Supplier Medchemexpress LLC Supplier No. HY1063425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMG 517 is a potent, selective small-molecule antagonist of the vanilloid receptor TRPV1 for research use. It inhibits TRPV1-mediated calcium influx in cell-based and neuronal assays with low-nanomolar IC50 values, and is supplied as a high-purity solid suitable for pharmacology and biochemical studies.

  • High purity solid suitable for in vitro research.
  • Low-nanomolar TRPV1 potency (IC50 ≈ 0.5-0.9 nM).
  • Molecular weight 430.40, formula C20H13F3N4O2S.
  • Supplied with data sheet, certificate of analysis, and safety data sheet.
  • Soluble in DMSO for preparation of stock solutions.
  • Available in multiple pack sizes for flexibility in study scale.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.