Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(4,6-dimethyl-2-pyridinyl)-4-[[4-(trifluoromethyl)phenyl]methyl]piperazine-1-carbothioamide | 1916571-90-8 | 99.7% | 408.48 g/mol | C20H23F3N4S | 1 ML
SDP

Catalog No. 5000248717
Encompass_Preferred

NCT-503 is a small-molecule inhibitor of phosphoglycerate dehydrogenase (PHGDH) used in biochemical and cellular research. It exhibits an in vitro IC50 of 2.5 μM and is provided as either a solid or a ready-to-use 10 mM solution in DMSO for cell-based and biochemical assays.

  • Inhibits PHGDH with an IC50 of 2.5 μM.
  • High purity (99.69%).
  • Molecular weight 408.48 g/mol.
  • Available as a 10 mM DMSO solution (1 mL) and multiple solid sizes.
  • Soluble in DMSO (50 mg/mL) and ethanol (13.33 mg/mL) with recommended handling.
  • Recommended storage: powder at -20°C (long term) or 4°C; solutions at -80°C or -20°C.
  • Intended for research use only.

Catalog No. 50-002-48717 Supplier Medchemexpress LLC Supplier No. HY10196610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NCT-503 is a small-molecule inhibitor of phosphoglycerate dehydrogenase (PHGDH) used in biochemical and cellular research. It exhibits an in vitro IC50 of 2.5 μM and is provided as either a solid or a ready-to-use 10 mM solution in DMSO for cell-based and biochemical assays.

  • Inhibits PHGDH with an IC50 of 2.5 μM.
  • High purity (99.69%).
  • Molecular weight 408.48 g/mol.
  • Available as a 10 mM DMSO solution (1 mL) and multiple solid sizes.
  • Soluble in DMSO (50 mg/mL) and ethanol (13.33 mg/mL) with recommended handling.
  • Recommended storage: powder at -20°C (long term) or 4°C; solutions at -80°C or -20°C.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.