Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Amino-6-ethoxy-1,3,5-triazin-2-yl)-N-methylamine, 97%, Thermo Scientific™

Catalog No. AC35726DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC35726DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC35726DA Supplier Thermo Scientific Chemicals Supplier No. AC35726DA
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62096-63-3
Molecular Formula C6H11N5O
Molecular Weight (g/mol) 169.188
InChI Key HEMSJLNVJJUYEU-UHFFFAOYSA-N
Synonym 6-ethoxy-n2-methyl-1,3,5-triazine-2,4-diamine, 2-amino-6-ethoxy-4-methylamino-1,3,5-triazine, 2-amino-4-methylamino-6-ethoxy-1,3,5-triazine, n-4-amino-6-ethoxy-1,3,5-triazin-2-yl-n-methylamine, 2-amino-4-methylamino-6-ethoxy-1,3,5-trazine, 2-amino-4-ethoxy-6-methylamino-1,3,5-triazine, 4-amino-6-ethoxy 1,3,5-triazin-2-yl methylamine, maybridge3_000309, 6-ethoxy-n-methyl-1,3,5-triazine-2,4-diamine, 4-ethoxy-6-methylamino-1,3,5-triazin-2-amine
PubChem CID 693795
IUPAC Name 6-ethoxy-2-N-methyl-1,3,5-triazine-2,4-diamine
SMILES CCOC1=NC(=NC(=N1)NC)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 169.19
Chemical Name or Material N-(4-amino-6-ethoxy-1, 3, 5-triazin-2-yl)-N-methylamine
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.