Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(4-aminobenzoyl)-beta-alanine | 7377-08-4 | MFCD00009805 | 99.4% | 208.22 g·mol⁻1 | C10H12N2O3 | 100 G
SDP

Catalog No. 5000303945
Encompass_Preferred

N-(4-Aminobenzoyl)-beta-alanine is an alanine derivative provided as a high-purity research reagent for biochemical and pharmacological studies. Supplied as a solid, it is commonly used as a reference standard or in studies of amino acid metabolism and related assays.

  • White to off-white solid suitable for analytical use.
  • CAS number 7377-08-4.
  • Molecular weight 208.22 g·mol⁻1.
  • Chemical formula C10H12N2O3.
  • Purity 99.35%.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended; use newly opened DMSO.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months).
  • Available in multiple pack sizes including 100 g.

Catalog No. 50-003-03945 Supplier Medchemexpress LLC Supplier No. HYW014505100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

N-(4-Aminobenzoyl)-beta-alanine is an alanine derivative provided as a high-purity research reagent for biochemical and pharmacological studies. Supplied as a solid, it is commonly used as a reference standard or in studies of amino acid metabolism and related assays.

  • White to off-white solid suitable for analytical use.
  • CAS number 7377-08-4.
  • Molecular weight 208.22 g·mol⁻1.
  • Chemical formula C10H12N2O3.
  • Purity 99.35%.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended; use newly opened DMSO.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months).
  • Available in multiple pack sizes including 100 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.