Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Aminobenzoyl)-L-glutamic Acid Diethyl Ester 98.0+%, TCI America™
SDP

Catalog No. A124025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A124025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A124025G Supplier TCI America Supplier No. A124025G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13726-52-8
Molecular Formula C16H22N2O5
Molecular Weight (g/mol) 322.361
MDL Number MFCD00009617
InChI Key RJXFBLRRPYBPTM-ZDUSSCGKSA-N
Synonym n-4-aminobenzoyl-l-glutamic acid diethyl ester, diethyl n-4-aminobenzoyl-l-glutamate, diethyl n-p-aminobenzoyl-l-glutamate, 1,5-diethyl 2s-2-4-aminophenyl formamido pentanedioate, 4-abz-glu oet-oet, l-glutamic acid, n-4-aminobenzoyl-, diethyl ester, diethyl 4-aminobenzoyl-l-glutamate, diethyl p-aminobenzoyl-l-glutamate, s-diethyl 2-4-aminobenzamido pentanedioate, diethyl 2-4-aminobenzoyl amino pentanedioate #
PubChem CID 73002
IUPAC Name diethyl (2S)-2-[(4-aminobenzoyl)amino]pentanedioate
SMILES CCOC(=O)CCC(C(=O)OCC)NC(=O)C1=CC=C(C=C1)N

Specifications

Melting Point 144°C
Quantity 25 g
Formula Weight 322.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminobenzoyl)-L-glutamic Acid Diethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.