Learn More
Medchemexpress LLC N-(4-aminobutyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide TFA salt | 1799711-25-3 | MFCD31560484 | 99.9% | 516.42 g·mol⁻¹ | C21H23F3N4O8 | 2 G
SDP
Supplier:  Medchemexpress LLC HY1036132G
Encompass_Preferred
Thalidomide-O-amido-C4-NH2 TFA is a thalidomide-derived cereblon ligand-linker conjugate provided as the trifluoroacetate (TFA) salt for use in PROTAC design and other chemical biology applications. It is supplied as a white to off-white solid with high analytical purity and a molecular weight of 516.42 g·mol⁻¹.
  • Thalidomide-derived cereblon ligand-linker conjugate.
  • Provided as the trifluoroacetate (TFA) salt.
  • High purity by LCMS (reported 99.92%).
  • White to off-white solid appearance.
  • Molecular formula C21H23F3N4O8; molecular weight 516.42 g·mol⁻¹.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-45152