Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Aminobutyl)-2-nitrobenzenesulfonamide 95.0+%, TCI America™
SDP

Catalog No. A16301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16301G Supplier TCI America Supplier No. A16301G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 211512-13-9
Molecular Formula C10H15N3O4S
Molecular Weight (g/mol) 273.307
MDL Number MFCD04117909
InChI Key NEIRGPMZEKSCSV-UHFFFAOYSA-N
Synonym 1-Amino-4-(2-nitrobenzenesulfonamido)butane, N-(2-Nitrobenzenesulfonyl)-1,4-butanediamine, N-(o-Ns)-1,4-diaminobutane
PubChem CID 11323272
IUPAC Name N-(4-aminobutyl)-2-nitrobenzenesulfonamide
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)NCCCCN

Specifications

Quantity 1 g
Formula Weight 273.31
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminobutyl)-2-nitrobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.