Learn More
Medchemexpress LLC N-(4-aminobutyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide | 151294-96-1 | MFCD28397129 | 97.0% | 314.45 g/mol | C14H26N4O2S | 250 MG
SDP
Catalog No. 5000204024
Encompass_Preferred
Biotin-C4-amide-C5-NH2 is a biotinylated, cleavable antibody-drug conjugate (ADC) linker supplied as a solid for research use. It contains a primary amine spacer for conjugation and is used for affinity-based capture and ADC synthesis workflows.
  • Biotinylated cleavable linker for ADC synthesis.
  • Primary amine functional group for conjugation.
  • High purity, suitable for research applications.
  • Solid form with standard storage recommendations for small molecules.
Catalog No. 50-002-04024 Supplier Medchemexpress LLC Supplier No. HYW096148250MG
Biotin-C4-amide-C5-NH2 is a biotinylated, cleavable antibody-drug conjugate (ADC) linker supplied as a solid for research use. It contains a primary amine spacer for conjugation and is used for affinity-based capture and ADC synthesis workflows.
  • Biotinylated cleavable linker for ADC synthesis.
  • Primary amine functional group for conjugation.
  • High purity, suitable for research applications.
  • Solid form with standard storage recommendations for small molecules.