Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(4-aminobutyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide | 151294-96-1 | MFCD28397129 | 97.0% | 314.45 g/mol | C14H26N4O2S | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW09614850MG

Encompass_Preferred

Biotin-C4-amide-C5-NH2 is a cleavable antibody-drug conjugate (ADC) linker bearing a biotin tag and a primary amine for affinity capture and conjugation during ADC synthesis and analysis. Intended for research use only.

  • Cleavable ADC linker with biotin tag for affinity-based capture.
  • Primary amine enables conjugation to payloads or linkers.
  • High purity: 97.0% (typical).
  • Molecular weight 314.45 g/mol; formula C14H26N4O2S.
  • Storage: store at 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • For research use only; not for human therapeutic use.

Catalog No. 50-002-03872


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.