Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Aminobutyl)acetamide 98.0+%, TCI America™
SDP

Catalog No. A23925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23925G Supplier TCI America Supplier No. A23925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5699-41-2
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD10024834
InChI Key KLZGKIDSEJWEDW-UHFFFAOYSA-N
Synonym N-Acetyl-1,4-butanediamine
PubChem CID 122356
ChEBI CHEBI:17768
IUPAC Name N-(4-aminobutyl)acetamide
SMILES CC(=O)NCCCCN

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 g
UN Number 2735
Formula Weight 130.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(4-Aminobutyl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.