Learn More
Medchemexpress LLC N-[[4-(benzamidocarbamoyl)phenyl]methyl]-3-chloro-4-fluorobenzenesulfonamide | 852918-02-6 | MFCD06683895 | 99.2% | C21H17ClFN3O4S | 10MG
SDP
Catalog No. 5000260049
Encompass_Preferred
TCN 201 is a potent, selective, non-competitive antagonist of GluN1/GluN2A (NR2A-containing) NMDA receptors used as a research tool in neuroscience. It inhibits NR2A-containing receptors with a reported pIC50 ≈ 6.8 and shows markedly lower activity at NR2B-containing receptors. The compound is supplied at high purity and is intended for use in in vitro and in vivo experimental applications.
  • Selective antagonist of NR2A-containing NMDA receptors.
  • Potent activity (pIC50 ≈ 6.8 for NR2A).
  • High chemical purity suitable for research (99.2%).
  • Well characterized molecular properties (C21H17ClFN3O4S, MW 461.89).
  • Useful for electrophysiology, pharmacology, and receptor subtype studies.
Catalog No. 50-002-60049 Supplier Medchemexpress LLC Supplier No. HY1345710MG
Add to Cart
Add to Cart
TCN 201 is a potent, selective, non-competitive antagonist of GluN1/GluN2A (NR2A-containing) NMDA receptors used as a research tool in neuroscience. It inhibits NR2A-containing receptors with a reported pIC50 ≈ 6.8 and shows markedly lower activity at NR2B-containing receptors. The compound is supplied at high purity and is intended for use in in vitro and in vivo experimental applications.
  • Selective antagonist of NR2A-containing NMDA receptors.
  • Potent activity (pIC50 ≈ 6.8 for NR2A).
  • High chemical purity suitable for research (99.2%).
  • Well characterized molecular properties (C21H17ClFN3O4S, MW 461.89).
  • Useful for electrophysiology, pharmacology, and receptor subtype studies.