Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(4-benzyl-3-methyl-1,2,4-thiadiazol-5-ylidene)-N'-phenylbenzenecarboximidamide | 324579-65-9 | 99.9% | 384.50 g/mol | C23H20N4S | 50MG
SDP

Catalog No. 5000239079
Encompass_Preferred

RNF5 inhibitor inh-02 is a small-molecule inhibitor of the E3 ubiquitin ligase RNF5 used in biochemical and cellular research to modulate ubiquitination and to rescue F508del-CFTR function in model systems. The compound has defined physicochemical properties and high purity, and is supplied in mg-scale quantities suitable for preparation of concentrated DMSO stock solutions for in vitro assays.

  • Inhibits E3 ubiquitin ligase RNF5 for cellular and biochemical assays.
  • C23H20N4S; molecular weight 384.5 g/mol.
  • High purity (99.91%).
  • Soluble in DMSO (≈83.33 mg/mL) with ultrasonic/warming and heating to 60°C if needed.
  • Recommended storage: powder -20°C (up to 3 years); solution -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for use in patients.

Catalog No. 50-002-39079 Supplier Medchemexpress LLC Supplier No. HY12396750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RNF5 inhibitor inh-02 is a small-molecule inhibitor of the E3 ubiquitin ligase RNF5 used in biochemical and cellular research to modulate ubiquitination and to rescue F508del-CFTR function in model systems. The compound has defined physicochemical properties and high purity, and is supplied in mg-scale quantities suitable for preparation of concentrated DMSO stock solutions for in vitro assays.

  • Inhibits E3 ubiquitin ligase RNF5 for cellular and biochemical assays.
  • C23H20N4S; molecular weight 384.5 g/mol.
  • High purity (99.91%).
  • Soluble in DMSO (≈83.33 mg/mL) with ultrasonic/warming and heating to 60°C if needed.
  • Recommended storage: powder -20°C (up to 3 years); solution -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for use in patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.