Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Benzyloxytrityl)-N'-(2-nitrobenzenesulfonyl)-1,4-diaminobutane Resin cross-linked with 1% DVB, TCI America™
SDP

Catalog No. B23721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23721G Supplier TCI America Supplier No. B23721G
Add to Cart
Add to Cart

(200-400mesh) (0.9-1.1mmol/g)

Chemical Identifiers

Synonym 4-[[4-(2-Nitrobenzenesulfonamido)butylamino]diphenylmethyl]phenoxymethyl Polystyrene Resin

Specifications

Quantity 1 g
Chemical Name or Material N-(4-Benzyloxytrityl)-N′-(2-nitrobenzenesulfonyl)-1,4-diaminobutane Resin cross-linked with 1%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.