Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(4-Bromophenyl)-N-Phenylacrylamide

Catalog No. 911798100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-179-8100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-179-8100MG Supplier Sigma Aldrich Supplier No. 911798100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(4-Bromophenyl)-N-phenylacrylamide is a cysteine-reactive small-molecule fragment for chemoproteomic and ligandability studies for both traditionally druggable proteins as well as undruggable, or difficult-to-target, proteins. This fragment electrophile, or scout fragment, can be used alone in fragment-based covalent ligand discovery or incorporated into bifunctional tools such as electrophilic PROTAC™ molecules for targeted protein degradation as demonstrated by the Cravatt Lab for E3 ligase discovery.Technology Spotlight: Proteomic Ligandability AssessmentProteome-wide covalent ligand discovery in native biological systemsElectrophilic PROTACs that degrade nuclear proteins by engaging DCAF16PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 29463-06-7
Molecular Formula C16H23BBrNO4
Quantity 100 mg
Synonym Benzyl boronate tag; Bromoethyl boronic acid pinacol ester nuclear tag; Nucleus-targeting probe building block
Molecular Weight (g/mol) 384.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.