Learn More
Medchemexpress LLC N-(4-butylphenyl)-5-nitrofuran-2-carboxamide | 346691-38-1 | MFCD00784211 | 99.7% | 288.30 | C15H16N2O4 | 5 MG
SDP
Catalog No. 5000223412
Encompass_Preferred
STING-IN-2 is a potent, covalent inhibitor of the stimulator of interferon genes (STING) protein used as a research tool to study STING-mediated signaling, innate immune responses, and autoinflammatory disease mechanisms. It inhibits both human and mouse STING and is supplied in small milligram quantities for in vitro and preclinical research.
  • Potent covalent inhibition of STING.
  • Active against human and mouse STING.
  • Useful for studying STING-mediated innate immune signaling.
  • Suitable for in vitro and preclinical research applications.
  • Characterized and high-purity material for research use.
Catalog No. 50-002-23412 Supplier Medchemexpress LLC Supplier No. HY1386825MG
STING-IN-2 is a potent, covalent inhibitor of the stimulator of interferon genes (STING) protein used as a research tool to study STING-mediated signaling, innate immune responses, and autoinflammatory disease mechanisms. It inhibits both human and mouse STING and is supplied in small milligram quantities for in vitro and preclinical research.
  • Potent covalent inhibition of STING.
  • Active against human and mouse STING.
  • Useful for studying STING-mediated innate immune signaling.
  • Suitable for in vitro and preclinical research applications.
  • Characterized and high-purity material for research use.