Learn More
Medchemexpress LLC N-(4-chloro-3-nitrophenyl)-4-nitro-2,1,3-benzoxadiazol-5-amine | 313964-19-1 | 99.9% | 335.66 g/mol | C12H6ClN5O5 | 10 MG
SDP
Catalog No. 5000297933
Encompass_Preferred
HIF-2α-IN-3 is a small-molecule allosteric inhibitor of hypoxia-inducible factor-2α (HIF-2α) used as a research reagent in studies of hypoxia signaling and oncology. Reported biochemical activity includes an IC50 of 0.4 μM and a KD of 1.1 μM; the compound is supplied as a high-purity solid for laboratory use.
  • Allosteric HIF-2α inhibition with reported IC50 0.4 μM and KD 1.1 μM.
  • Suitable for in vitro studies of hypoxia signaling and cancer biology.
  • High purity suitable for research applications.
  • Supplied as a light yellow solid for standard laboratory handling.
Catalog No. 50-002-97933 Supplier Medchemexpress LLC Supplier No. HY1837010MG
HIF-2α-IN-3 is a small-molecule allosteric inhibitor of hypoxia-inducible factor-2α (HIF-2α) used as a research reagent in studies of hypoxia signaling and oncology. Reported biochemical activity includes an IC50 of 0.4 μM and a KD of 1.1 μM; the compound is supplied as a high-purity solid for laboratory use.
  • Allosteric HIF-2α inhibition with reported IC50 0.4 μM and KD 1.1 μM.
  • Suitable for in vitro studies of hypoxia signaling and cancer biology.
  • High purity suitable for research applications.
  • Supplied as a light yellow solid for standard laboratory handling.