Learn More
Medchemexpress LLC N-(4'-cyano[1,1'-biphenyl]-4-yl)-N'-[4-(3H-imidazo[4,5-b]pyridin-7-yl)phenyl]urea | 1361414-26-7 | 98.1% | 25 MG
SDP
Catalog No. 5000234865
Encompass_Preferred
SR9186 (ML368) is a selective CYP3A4 inhibitor research compound used to study CYP3A4-mediated metabolism and hepatic-stage Plasmodium falciparum. It exhibits low-nanomolar inhibitory potency against common CYP3A4 substrates and is supplied as a high-purity powder for laboratory research.
  • Selective CYP3A4 inhibitor with low-nanomolar potency.
  • High purity (98.1%).
  • Molecular formula C26H18N6O; molecular weight 430.46 g/mol.
  • Stable as a powder under recommended cold storage conditions.
  • Suitable for in vitro metabolism and pharmacology studies.
Catalog No. 50-002-34865 Supplier Medchemexpress LLC Supplier No. HY12069625MG
SR9186 (ML368) is a selective CYP3A4 inhibitor research compound used to study CYP3A4-mediated metabolism and hepatic-stage Plasmodium falciparum. It exhibits low-nanomolar inhibitory potency against common CYP3A4 substrates and is supplied as a high-purity powder for laboratory research.
  • Selective CYP3A4 inhibitor with low-nanomolar potency.
  • High purity (98.1%).
  • Molecular formula C26H18N6O; molecular weight 430.46 g/mol.
  • Stable as a powder under recommended cold storage conditions.
  • Suitable for in vitro metabolism and pharmacology studies.