Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(4'-cyano[1,1'-biphenyl]-4-yl)-N'-[4-(3H-imidazo[4,5-b]pyridin-7-yl)phenyl]urea | 1361414-26-7 | 98.1% | 50 MG
SDP

Catalog No. 5000232791
Encompass_Preferred

SR9186 (ML368) is a research-grade small-molecule inhibitor of cytochrome P450 3A4 (CYP3A4), supplied for in vitro and preclinical research use. It is available as a powder and as a DMSO solution for assay preparation.

  • Selective CYP3A4 inhibition with low-nanomolar activity for common substrates
  • High purity suitable for biochemical and cell-based assays
  • Available as powder and as a 10 mM solution in DMSO for convenient dosing
  • Molecular weight ~430.46 and formula C26H18N6O
  • Recommended storage: powder at -20°C (long-term) or 4°C (short-term); solution at -80°C

Catalog No. 50-002-32791 Supplier Medchemexpress LLC Supplier No. HY12069650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

SR9186 (ML368) is a research-grade small-molecule inhibitor of cytochrome P450 3A4 (CYP3A4), supplied for in vitro and preclinical research use. It is available as a powder and as a DMSO solution for assay preparation.

  • Selective CYP3A4 inhibition with low-nanomolar activity for common substrates
  • High purity suitable for biochemical and cell-based assays
  • Available as powder and as a 10 mM solution in DMSO for convenient dosing
  • Molecular weight ~430.46 and formula C26H18N6O
  • Recommended storage: powder at -20°C (long-term) or 4°C (short-term); solution at -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.