Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(4-cyano-3-(trifluoromethyl)phenyl)-3-((3-fluorophenyl)sulfinyl)-2-hydroxy-2-methylpropanamide | 1539314-06-1 | MFCD30343844 | 99.4% | 330.33 g/mol | C14H13F3N2O2S | 50 MG
SDP

Catalog No. 5000215223
Encompass_Preferred

GSK-2881078 is an orally active, nonsteroidal selective androgen receptor modulator (SARM) used for preclinical research into muscle wasting and cachexia. It shows tissue-selective AR activity and induces AR-mediated transcriptional activation in cellular assays (PC3(AR)2 EC50 = 3.99 nM).

  • Orally active, nonsteroidal SARM suitable for in vitro and preclinical studies.
  • Tissue-selective AR modulation with potent receptor activation (EC50 = 3.99 nM in PC3(AR)2 cells).
  • High reported purity of 99.42% (supplier data).
  • Molecular formula C14H13F3N2O2S; molecular weight 330.33 g/mol.
  • Available as solid and as 10 mM solution in DMSO for convenience in assays.
  • CAS number 1539314-06-1 provided for unambiguous identification.

Catalog No. 50-002-15223 Supplier Medchemexpress LLC Supplier No. HY10018650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK-2881078 is an orally active, nonsteroidal selective androgen receptor modulator (SARM) used for preclinical research into muscle wasting and cachexia. It shows tissue-selective AR activity and induces AR-mediated transcriptional activation in cellular assays (PC3(AR)2 EC50 = 3.99 nM).

  • Orally active, nonsteroidal SARM suitable for in vitro and preclinical studies.
  • Tissue-selective AR modulation with potent receptor activation (EC50 = 3.99 nM in PC3(AR)2 cells).
  • High reported purity of 99.42% (supplier data).
  • Molecular formula C14H13F3N2O2S; molecular weight 330.33 g/mol.
  • Available as solid and as 10 mM solution in DMSO for convenience in assays.
  • CAS number 1539314-06-1 provided for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.