Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide 98.0+%, TCI America™
SDP

Catalog No. C31881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31881G 1 g
C31885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31881G Supplier TCI America Supplier No. C31881G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90357-53-2
Molecular Formula C12H9F3N2O
Molecular Weight (g/mol) 254.212
MDL Number MFCD03411609
InChI Key HHWDZLSGDDXUSM-UHFFFAOYSA-N
PubChem CID 11149469
IUPAC Name N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
SMILES CC(=C)C(=O)NC1=CC(=C(C=C1)C#N)C(F)(F)F

Specifications

Melting Point 143°C
Color White
Quantity 1 g
UN Number 3077
Formula Weight 254.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.