Learn More
Medchemexpress LLC N'-(4-(diethylamino)-2-hydroxybenzylidene)isonicotinohydrazide | 140405-36-3 | MFCD00542031 | 99.7% | 312.37 | C17H20N4O2 | 100 MG
SDP
Catalog No. 5000246124
Encompass_Preferred
RSVA405 is a small-molecule activator of AMP-activated protein kinase (AMPK) that promotes CaMKKβ-dependent AMPK activation, inhibits mTOR signaling, enhances autophagy and Aβ degradation, and exhibits anti-inflammatory activity via STAT3 inhibition. It is used in metabolic, neurodegenerative, and obesity research.
  • Potent AMPK activation and mTOR inhibition.
  • Promotes autophagy and Aβ degradation.
  • Reported anti-inflammatory activity through STAT3 inhibition.
  • High purity (99.72%) suitable for research applications.
  • Available as solid and as 10 mM solution in DMSO; multiple package sizes.
  • Stable when stored as powder at -20°C and in solvent under recommended conditions.
Catalog No. 50-002-46124 Supplier Medchemexpress LLC Supplier No. HY103238100MG
RSVA405 is a small-molecule activator of AMP-activated protein kinase (AMPK) that promotes CaMKKβ-dependent AMPK activation, inhibits mTOR signaling, enhances autophagy and Aβ degradation, and exhibits anti-inflammatory activity via STAT3 inhibition. It is used in metabolic, neurodegenerative, and obesity research.
  • Potent AMPK activation and mTOR inhibition.
  • Promotes autophagy and Aβ degradation.
  • Reported anti-inflammatory activity through STAT3 inhibition.
  • High purity (99.72%) suitable for research applications.
  • Available as solid and as 10 mM solution in DMSO; multiple package sizes.
  • Stable when stored as powder at -20°C and in solvent under recommended conditions.