Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N'-(4-(diethylamino)-2-hydroxybenzylidene)isonicotinohydrazide | 140405-36-3 | MFCD00542031 | 99.7% | 312.37 g/mol | C17H20N4O2 | 10 MG
SDP

Catalog No. 5000245937
Encompass_Preferred

RSVA405 (CAS 140405-36-3) is a small-molecule activator of AMP-activated protein kinase (AMPK) used in cellular and biochemical research. It promotes CaMKKβ-dependent AMPK activation, inhibits mTOR signaling, and can induce autophagy; supplied at high purity for research applications.

  • Potent AMPK activator; reported EC50 = 1 μM.
  • High purity (~99.7%).
  • Molecular formula C17H20N4O2; molecular weight 312.37 g/mol.
  • Soluble in DMSO; available as solid or DMSO solution.
  • For research use only; not for human consumption.

Catalog No. 50-002-45937 Supplier Medchemexpress LLC Supplier No. HY10323810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RSVA405 (CAS 140405-36-3) is a small-molecule activator of AMP-activated protein kinase (AMPK) used in cellular and biochemical research. It promotes CaMKKβ-dependent AMPK activation, inhibits mTOR signaling, and can induce autophagy; supplied at high purity for research applications.

  • Potent AMPK activator; reported EC50 = 1 μM.
  • High purity (~99.7%).
  • Molecular formula C17H20N4O2; molecular weight 312.37 g/mol.
  • Soluble in DMSO; available as solid or DMSO solution.
  • For research use only; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.