Learn More
Medchemexpress LLC N-(4-(difluoromethoxy)phenyl)-2-((6-oxo-4-propyl-1,6-dihydropyrimidin-2-yl)thio)acetamide | 502887-71-0 | 99.8% | C16H17F2N3O3S | 10MG
SDP
Catalog No. 5000260777
Encompass_Preferred
MMP-9-IN-1 is a selective inhibitor of matrix metalloproteinase-9 (MMP-9) that binds the hemopexin (PEX) domain and shows minimal activity against other MMP family members. Intended as a research reagent, the compound is described with formula C16H17F2N3O3S, molecular weight 369.39, and a reported high purity for biochemical and cellular studies.
  • Selectively targets the hemopexin (PEX) domain of MMP-9.
  • High chemical purity suitable for research applications.
  • Characterized molecular formula and molecular weight for reference.
  • Available in multiple vial sizes to support assay scaling.
  • Provided with SMILES and structural data for compound identification.
Catalog No. 50-002-60777 Supplier Medchemexpress LLC Supplier No. HY13523210MG
MMP-9-IN-1 is a selective inhibitor of matrix metalloproteinase-9 (MMP-9) that binds the hemopexin (PEX) domain and shows minimal activity against other MMP family members. Intended as a research reagent, the compound is described with formula C16H17F2N3O3S, molecular weight 369.39, and a reported high purity for biochemical and cellular studies.
  • Selectively targets the hemopexin (PEX) domain of MMP-9.
  • High chemical purity suitable for research applications.
  • Characterized molecular formula and molecular weight for reference.
  • Available in multiple vial sizes to support assay scaling.
  • Provided with SMILES and structural data for compound identification.