Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,4'-Dimethylformanilide 97.0+%, TCI America™
SDP

Catalog No. D33691G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33691G 1 g
D33695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33691G Supplier TCI America Supplier No. D33691G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2739-04-0
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
InChI Key MQMOWSBDSHOJAV-UHFFFAOYSA-N
Synonym N-Methyl-N-(p-tolyl)formamide
PubChem CID 75957
IUPAC Name N-methyl-N-(4-methylphenyl)formamide
SMILES CC1=CC=C(C=C1)N(C)C=O

Specifications

Color Yellow
Boiling Point 277°C
Quantity 1 g
Formula Weight 149.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,4′-Dimethylformanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.