Learn More
Medchemexpress LLC N-[4-[(E)-2-[3-t-Bu-5-(2,4-dioxopyrimidin-1-yl)-2-methoxyphenyl]ethenyl]phenyl]methanesulfonamide | 1132936-00-5 | MFCD28502074 | 99.8% | 469.55 g/mol | C24H27N3O5S | 25 MG
SDP
Catalog No. 5000248506
Encompass_Preferred
ABT-072 is an orally active, non-nucleoside inhibitor of the hepatitis C virus NS5B RNA-dependent RNA polymerase. It exhibits nanomolar in vitro potency against genotype 1a and 1b polymerases and showed favorable pharmacokinetics in preclinical studies, including low plasma clearance and good oral bioavailability.
  • Orally active non-nucleoside NS5B polymerase inhibitor.
  • Nanomolar potency against HCV genotype 1a and 1b (EC50 values reported).
  • Favorable pharmacokinetic profile in preclinical species.
  • High chemical purity suitable for research applications.
  • Solid, off-white to yellow appearance.
  • Stable under recommended cold storage conditions.
Catalog No. 50-002-48506 Supplier Medchemexpress LLC Supplier No. HY10163425MG
ABT-072 is an orally active, non-nucleoside inhibitor of the hepatitis C virus NS5B RNA-dependent RNA polymerase. It exhibits nanomolar in vitro potency against genotype 1a and 1b polymerases and showed favorable pharmacokinetics in preclinical studies, including low plasma clearance and good oral bioavailability.
  • Orally active non-nucleoside NS5B polymerase inhibitor.
  • Nanomolar potency against HCV genotype 1a and 1b (EC50 values reported).
  • Favorable pharmacokinetic profile in preclinical species.
  • High chemical purity suitable for research applications.
  • Solid, off-white to yellow appearance.
  • Stable under recommended cold storage conditions.