Learn More
Medchemexpress LLC N-[4-[(E)-2-[3-t-Bu-5-(2,4-dioxopyrimidin-1-yl)-2-methoxyphenyl]ethenyl]phenyl]methanesulfonamide | 1132936-00-5 | MFCD28502074 | 99.8% | 469.6 g/mol | C24H27N3O5S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10163450MG
Encompass_Preferred
ABT-072 is an orally active, non-nucleoside inhibitor of the hepatitis C virus NS5B polymerase used as a research compound. It demonstrates sub-nanomolar to low-nanomolar antiviral activity against genotype 1 polymerases and is supplied for in vitro and preclinical investigations.
  • Potent NS5B inhibition (GT1a EC50 = 1 nM; GT1b EC50 = 0.3 nM).
  • Orally bioavailable profile observed in preclinical studies.
  • High purity suitable for research use.
  • Well-characterized chemical identity (CAS 1132936-00-5).
  • Suitable as a reference compound in antiviral assays.
Catalog No. 50-002-45340