Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Ethoxybenzylidene)-4-acetylaniline 98.0+%, TCI America™
SDP

Catalog No. E02541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E02541G Supplier TCI America Supplier No. E02541G

Chemical Identifiers

CAS 17224-17-8
Molecular Formula C17H17NO2
Molecular Weight (g/mol) 267.328
MDL Number MFCD00059374
InChI Key HCDYNBNSFSMUIM-UHFFFAOYSA-N
Synonym 4′C-Ethoxybenzylidene-4-aminoacetophenone, 4′C-Ethoxybenzylidene-4-acetylaniline, 4′C-[(4-Ethoxybenzylidene)amino]acetophenone
PubChem CID 40424582
IUPAC Name 1-[4-[(4-ethoxyphenyl)methylideneamino]phenyl]ethanone
SMILES CCOC1=CC=C(C=C1)C=NC2=CC=C(C=C2)C(=O)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 267.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Ethoxybenzylidene)-4-acetylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.