Learn More
Medchemexpress LLC N-(4-ethynylphenyl)-5,7-dimethyl-pyrazolo[1,5-a]pyrimidine-3-carboxamide | 1380716-06-2 | 98.0% | 290.32 g·mol⁻¹ | C17H14N4O | 25 MG
SDP
Catalog No. 5000227908
Encompass_Preferred
ML198 is a small-molecule glucocerebrosidase (GCase) activator and non-inhibitory chaperone for research use. It has an EC50 of 0.4 μM and includes an alkyne functional group for click-chemistry, making it suitable for biochemical studies and probe development.
  • Activates glucocerebrosidase with EC50 0.4 μM.
  • Non-inhibitory chaperone activity.
  • Contains an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • High purity (≈98.0%).
  • Supplied in small research pack sizes for laboratory use.
Catalog No. 50-002-27908 Supplier Medchemexpress LLC Supplier No. HY11786125MG
ML198 is a small-molecule glucocerebrosidase (GCase) activator and non-inhibitory chaperone for research use. It has an EC50 of 0.4 μM and includes an alkyne functional group for click-chemistry, making it suitable for biochemical studies and probe development.
  • Activates glucocerebrosidase with EC50 0.4 μM.
  • Non-inhibitory chaperone activity.
  • Contains an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • High purity (≈98.0%).
  • Supplied in small research pack sizes for laboratory use.