Learn More
Medchemexpress LLC N-(4-formylphenyl)acetamide | 122-85-0 | MFCD00003380 | 99.7% | 163.17 g/mol | C9H9NO2 | 100 G
SDP
Catalog No. 5000210419
Encompass_Preferred
N-(4-formylphenyl)acetamide (4-acetamidobenzaldehyde) is an organic compound used as a biochemical assay reagent and intermediate in chemical synthesis. It is supplied as a high-purity solid for laboratory research and is suitable for use in biochemical assays and synthetic applications.
  • High purity: 99.7%.
  • Solid form for easy weighing and handling.
  • Available in multiple pack sizes, including 100 G.
  • Storage: keep at 4°C under nitrogen; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Identified by CAS 122-85-0 and formula C9H9NO2.
Catalog No. 50-002-10419 Supplier Medchemexpress LLC Supplier No. HYW067445100G
N-(4-formylphenyl)acetamide (4-acetamidobenzaldehyde) is an organic compound used as a biochemical assay reagent and intermediate in chemical synthesis. It is supplied as a high-purity solid for laboratory research and is suitable for use in biochemical assays and synthetic applications.
  • High purity: 99.7%.
  • Solid form for easy weighing and handling.
  • Available in multiple pack sizes, including 100 G.
  • Storage: keep at 4°C under nitrogen; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Identified by CAS 122-85-0 and formula C9H9NO2.