Learn More
Medchemexpress LLC N-(4-hydroxyphenyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide | 901770-40-9 | MFCD29037927 | 99.5% | C16H21N3O3S | 10MG
SDP
Catalog No. 5000261035
Encompass_Preferred
Biotin-4-aminophenol is a biotin-phenol analog used to generate biotinylated protein radicals for selective labeling of tyrosine residues in proteins. Supplied as a solid or as pre-dissolved 10 mM solutions in DMSO, it is intended for biochemical labeling and proteomics workflows where site-selective modification of proteins is required.
  • Selective tyrosine labeling via radical-mediated conjugation.
  • Efficient generation of biotinylated protein radicals for detection and enrichment.
  • Available as solid or 10 mM solution in DMSO for experimental convenience.
  • High purity suitable for biochemical and proteomics applications.
  • White to off-white solid with good solubility in DMSO.
Catalog No. 50-002-61035 Supplier Medchemexpress LLC Supplier No. HY14189810MG
Biotin-4-aminophenol is a biotin-phenol analog used to generate biotinylated protein radicals for selective labeling of tyrosine residues in proteins. Supplied as a solid or as pre-dissolved 10 mM solutions in DMSO, it is intended for biochemical labeling and proteomics workflows where site-selective modification of proteins is required.
  • Selective tyrosine labeling via radical-mediated conjugation.
  • Efficient generation of biotinylated protein radicals for detection and enrichment.
  • Available as solid or 10 mM solution in DMSO for experimental convenience.
  • High purity suitable for biochemical and proteomics applications.
  • White to off-white solid with good solubility in DMSO.