Learn More
Medchemexpress LLC N-(4-hydroxyphenyl)glycine | 122-87-2 | MFCD00002689 | 97.0% | 167.16 g/mol | C8H9NO3 | 1 G
SDP
Catalog No. 5000283050
Encompass_Preferred
N-(4-Hydroxyphenyl)glycine is a small-molecule amino acid derivative used for research and analytical applications. It is the para-hydroxyphenyl substituted glycine with molecular formula C8H9NO3, molecular weight 167.16 g/mol, and CAS number 122-87-2. Supplied as a light yellow to brown solid with reported purity ~97% and typically packaged in 1 g quantities for laboratory use.
  • Useful as an analytical standard or synthetic intermediate.
  • Reported high purity suitable for analytical workflows.
  • Solid form provides convenient storage and handling.
  • Compatible with common analytical techniques such as HPLC and mass spectrometry.
  • Supplied in research-scale quantities for laboratory use.
Catalog No. 50-002-83050 Supplier Medchemexpress LLC Supplier No. HYW1418851G
N-(4-Hydroxyphenyl)glycine is a small-molecule amino acid derivative used for research and analytical applications. It is the para-hydroxyphenyl substituted glycine with molecular formula C8H9NO3, molecular weight 167.16 g/mol, and CAS number 122-87-2. Supplied as a light yellow to brown solid with reported purity ~97% and typically packaged in 1 g quantities for laboratory use.
  • Useful as an analytical standard or synthetic intermediate.
  • Reported high purity suitable for analytical workflows.
  • Solid form provides convenient storage and handling.
  • Compatible with common analytical techniques such as HPLC and mass spectrometry.
  • Supplied in research-scale quantities for laboratory use.