Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(4-Iodo-2-methoxypyridin-3-yl)pivalamide

Catalog No. ADE0006241G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0006241G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0006241G Supplier Sigma Aldrich Supplier No. ADE0006241G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl 4-iodobenzoate can be used in the synthesis of biphenyl derivatives via negishi cross coupling with the intermediates of the type armg(tmp)-+2licl (tmp = trimethylolpropane)
  • It can also be used as a starting material in the synthesis of p-carbomethoxyphenylmagnesium bromide, a polyfunctional organometallic reagent that can be used as building block in the synthesis of complex target molecules

Specifications

CAS 131653-62-8
Molecular Formula C11H15IN2O2
Linear Formula C11H15IN2O2
MDL Number MFCD12026762
Quantity 5 g
Molecular Weight (g/mol) 334.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.