Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-4-Methanesulfonamidephenylboronic Acid

Catalog No. 7181811G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-818-11G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 71-818-11G Supplier Sigma Aldrich Supplier No. 7181811G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Potent and selective inhibitors of IKK-+aInhibitors for treatment of osteoporosisInhibitors of human farnesyl pyrophosphate synthaseSuzuki-type Pd(0) coupling reactions in the synthesis of 2-arylpurines as Cdk inhibitors

Specifications

CAS 756525-99-2
Molecular Formula C22H31N3O11
Linear Formula C22H31N3O11
MDL Number MFCD11041113
Quantity 100 mg
Synonym N-[15-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-15-oxo-3,6,9,12-tetraoxapentadec-1-yl]-2,5-dihydro-2,5-dioxo-1H-pyrrole-1-propanamide; monodisperse Mal-PEG4-NHS-ester
Molecular Weight (g/mol) 513.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.