Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[4-methoxy-3-(4-methyl-1-piperazinyl)phenyl]-3-methyl-4-(4-pyridinyl)benzamide | 148547-33-5 | 99.8% | 1 ML
SDP

Catalog No. 5000231054
Encompass_Preferred

GR 125743 (CAS 148547-33-5) is a selective 5-HT1B/1D receptor antagonist supplied as a research reagent. It is commonly provided as a 10 mM solution in DMSO or as solid quantities for preclinical pharmacology studies in neurological and cardiovascular research.

  • Selective 5-HT1B/1D receptor antagonist for receptor pharmacology studies.
  • Available as 10 mM solution in DMSO and in solid mg quantities.
  • High analytical purity (99.8%), suitable for research applications.
  • Molecular weight 416.52 g/mol; formula C25H28N4O2.

Catalog No. 50-002-31054 Supplier Medchemexpress LLC Supplier No. HY12139210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GR 125743 (CAS 148547-33-5) is a selective 5-HT1B/1D receptor antagonist supplied as a research reagent. It is commonly provided as a 10 mM solution in DMSO or as solid quantities for preclinical pharmacology studies in neurological and cardiovascular research.

  • Selective 5-HT1B/1D receptor antagonist for receptor pharmacology studies.
  • Available as 10 mM solution in DMSO and in solid mg quantities.
  • High analytical purity (99.8%), suitable for research applications.
  • Molecular weight 416.52 g/mol; formula C25H28N4O2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.