Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(4-Methoxybenzylidene)-4-butylaniline

Catalog No. 1582245G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-822-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-822-45G Supplier Sigma Aldrich Supplier No. 1582245G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(4-Methoxybenzylidene)-4-butylaniline (MBBA) is a nematic liquid crystal molecule that can be prepared by condensing distilled n-butylaniline and methoxybenzaldehyde. It can be used as an anisotropic solvent which has an isotropic transition temperature of 315k. It is majorly used in the development of liquid crystal cells.

Specifications

CAS 26227-73-6
Density 1.027 g/mL (at 25°C)
Molecular Formula C18H21NO
Linear Formula CH3(CH2)3C6H4N=CHC6H4OCH3
Refractive Index n20/D 1.5496 (literature)
MDL Number MFCD00009464
Quantity 5 g
Synonym MBBA
Molecular Weight (g/mol) 267.37
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.