Learn More
Medchemexpress LLC N-(4-methoxyphenyl)-1-phenyl-1H-pyrazol-3-amine | 103141-09-9 | MFCD00877802 | 99.3% | 265.31 | C16H15N3O | 50 MG
SDP
Catalog No. 5000242615
Encompass_Preferred
FPL 62064 is a small-molecule dual inhibitor of 5-lipoxygenase (5-LOX) and cyclooxygenase (COX) with demonstrated anti-inflammatory activity in vitro and in vivo. It exhibits low-micromolar inhibition of 5-LOX and COX in biochemical assays and reduces leukotriene and prostaglandin formation in cellular and animal models. Supplied as a solid with high reported purity for research use.
  • Potent 5-LOX and COX inhibition in biochemical assays.
  • Demonstrated anti-inflammatory activity in cellular and animal studies.
  • High reported purity suitable for research applications.
  • Soluble in DMSO at high concentration for in vitro experiments.
  • Stable under recommended storage conditions for solids and solutions.
Catalog No. 50-002-42615 Supplier Medchemexpress LLC Supplier No. HY10502450MG
FPL 62064 is a small-molecule dual inhibitor of 5-lipoxygenase (5-LOX) and cyclooxygenase (COX) with demonstrated anti-inflammatory activity in vitro and in vivo. It exhibits low-micromolar inhibition of 5-LOX and COX in biochemical assays and reduces leukotriene and prostaglandin formation in cellular and animal models. Supplied as a solid with high reported purity for research use.
  • Potent 5-LOX and COX inhibition in biochemical assays.
  • Demonstrated anti-inflammatory activity in cellular and animal studies.
  • High reported purity suitable for research applications.
  • Soluble in DMSO at high concentration for in vitro experiments.
  • Stable under recommended storage conditions for solids and solutions.