Learn More
Medchemexpress LLC N- 4-Methoxyphenyl m 1g
SDP
Catalog No. 5000204731
Encompass_Preferred
N-(4-Methoxyphenyl)maleimide (N-(p-Methoxyphenyl)maleimide) is a compound that can copolymerize with Ethyl acrylate[1][2]
Catalog No. 50-002-04731 Supplier Medchemexpress LLC Supplier No. HYW0977341G
N-(4-Methoxyphenyl)maleimide (N-(p-Methoxyphenyl)maleimide) is a compound that can copolymerize with Ethyl acrylate[1][2]