Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Methoxyphenyl)-p-phenylenediamine Hydrochloride 95%, Thermo Scientific™

Catalog No. AC413890050
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC413890050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC413890050 Supplier Thermo Scientific Chemicals Supplier No. 413890050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3566-44-7
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C13H15ClN2O
Quantity 5g
MDL Number MFCD00012989
Beilstein 13, III, 1161
Merck Index 15, 6072
Synonym variamine blue b, unii-e365pn4l70, n-4-methoxyphenyl-1,4-benzenediamine hydrochloride, n-4-methoxyphenyl benzene-1,4-diamine monohydrochloride, 1,4-benzenediamine, n-4-methoxyphenyl-, monohydrochloride, n-4-methoxyphenyl-p-phenylenediamine hydrochloride, 1,4-benzenediamine, n1-4-methoxyphenyl-, hydrochloride 1:1, 4-amino-4'-methoxydiphenylamine hydrochloride, n-p-methoxyphenyl-p-phenylenediamine hydrochloride, 4-aminophenyl 4-methoxyphenyl amine, chloride
InChI Key HPQQXLXIGHOKNZ-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.Cl
IUPAC Name 4-N-(4-methoxyphenyl)benzene-1,4-diamine;hydrochloride
Molecular Weight (g/mol) 250.726
PubChem CID 77108
Formula Weight 250.73
Percent Purity ≥94%
Chemical Name or Material N-(4-Methoxyphenyl)-p-phenylenediamine hydrochloride, 95%
Show More Show Less
EINECSNumber 222-652-7
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.