Learn More
Medchemexpress LLC N-[[4-methyl-2-(2-thienyl)-5-thiazolyl]methyl]-N′-2-thienyl-urea | 1396814-79-1 | 98.5% | 335.47 g/mol | C14H13N3OS3 | 10 MG
SDP
Catalog No. 5000288519
Encompass_Preferred
VU0542270 is a small-molecule research compound that selectively inhibits vascular Kir6.1/SUR2B ATP-sensitive potassium (KATP) channels and is used in cellular and biochemical assays to study SUR2B/Kir6.1-mediated KATP channel function. Reported potency is in the low-hundreds of nanomolar range, and the supplied sample is high-purity solid for laboratory research.
  • Selective inhibitor of vascular Kir6.1/SUR2B KATP channels.
  • Reported potency: IC50 ≈ 0.10-0.13 μM.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as a 10 mg solid research sample.
  • Commonly used in pharmacological and mechanistic studies of KATP channels.
Catalog No. 50-002-88519 Supplier Medchemexpress LLC Supplier No. HY16232210MG
VU0542270 is a small-molecule research compound that selectively inhibits vascular Kir6.1/SUR2B ATP-sensitive potassium (KATP) channels and is used in cellular and biochemical assays to study SUR2B/Kir6.1-mediated KATP channel function. Reported potency is in the low-hundreds of nanomolar range, and the supplied sample is high-purity solid for laboratory research.
  • Selective inhibitor of vascular Kir6.1/SUR2B KATP channels.
  • Reported potency: IC50 ≈ 0.10-0.13 μM.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as a 10 mg solid research sample.
  • Commonly used in pharmacological and mechanistic studies of KATP channels.